Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Piperidineethanol, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
3040-44-6
C7H15NO
129.2
MFCD00006512
KZTWONRVIPPDKH-UHFFFAOYSA-N
2-piperidinoethanol,1-piperidineethanol,2-piperidin-1-yl ethanol,1-2-hydroxyethyl piperidine,n-2-hydroxyethyl piperidine,n-piperidineethanol,2-1-piperidinyl ethanol,n-hydroxyethyl piperidine,beta-piperidylethanol,beta-piperidinoethanol
18232
CHEBI:61238
2-piperidin-1-ylethanol
C1CCN(CC1)CCO
3040-44-6
C7H15NO
129.2
MFCD00006512
KZTWONRVIPPDKH-UHFFFAOYSA-N
2-piperidinoethanol,1-piperidineethanol,2-piperidin-1-yl ethanol,1-2-hydroxyethyl piperidine,n-2-hydroxyethyl piperidine,n-piperidineethanol,2-1-piperidinyl ethanol,n-hydroxyethyl piperidine,beta-piperidylethanol,beta-piperidinoethanol
18232
CHEBI:61238
2-piperidin-1-ylethanol
C1CCN(CC1)CCO
Flash Point 77°C
Viscosity 20.9 mPa.s (20°C)
Specific Gravity 0.973
Melting Point 16.7°C
Boiling Point 198°C to 203°C
Refractive Index 1.4794 to 1.4814
Density 0.9730g/mL
Color Brown
Quantity 500g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Beilstein 20, 25
Solubility Information Solubility in water: miscible
Formula Weight 129.2
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material N-Piperidineethanol, 99%
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED

GHS Signal Word: Warning

EINECSNumber : 221-244-6

RTECSNumber : TM8052000

TSCA : TSCA