Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(Propargyloxy)phthalimide 98%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
25g
Packaging:
Glass bottle
4616-63-1
C11H7NO3
201.18
MFCD00005890
HBGZBVPXPDNXOV-UHFFFAOYSA-N
n-propargyloxy phthalimide,n-prop-2-ynyloxy phthalimide,2-prop-2-yn-1-yloxy isoindoline-1,3-dione,n-propargyloxyphthalimide,2-prop-2-yn-1-yloxy isoindole-1,3-dione,2-prop-2-ynyloxybenzo c azoline-1,3-dione,acmc-20aka8,n-2-propynoxyphthalimide,n-2-propynyloxy phthalimide,phthalimide, n-2-propynyloxy
78357
2-prop-2-ynoxyisoindole-1,3-dione
C#CCON1C(=O)C2=CC=CC=C2C1=O
4616-63-1
C11H7NO3
201.18
MFCD00005890
HBGZBVPXPDNXOV-UHFFFAOYSA-N
n-propargyloxy phthalimide,n-prop-2-ynyloxy phthalimide,2-prop-2-yn-1-yloxy isoindoline-1,3-dione,n-propargyloxyphthalimide,2-prop-2-yn-1-yloxy isoindole-1,3-dione,2-prop-2-ynyloxybenzo c azoline-1,3-dione,acmc-20aka8,n-2-propynoxyphthalimide,n-2-propynyloxy phthalimide,phthalimide, n-2-propynyloxy
78357
2-prop-2-ynoxyisoindole-1,3-dione
C#CCON1C(=O)C2=CC=CC=C2C1=O
Melting Point 149°C to 152°C
Color Beige to Brown
Quantity 10g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 201.18
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material N-(Propargyloxy)phthalimide, 98%
Show More Show Less

EINECSNumber : 225-024-0