Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ n-Propyl 3,4,5-trihydroxybenzoate, 98%

Manufacturer:  Alfa Aesar™ A108770E

Quantity 2500g

 View more versions of this product

Catalog No. AAA108770E


  Check Availability
Add to cart

Specifications

Description

Applications
n-Propyl 3,4,5-trihydroxybenzoate is an anti-fade reagent in fluorescence microscopy to reduce photobleaching of fluorescent probes such as rhodamine and fluorescein. Also used as an antioxidant.

Solubility
Soluble in ethanol (50 mg/ml) and water (0.35 g/100 ml at 25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Description & Specifications

Chemical Identifiers

121-79-9
212.2
1877976
Gallic acid n-propyl ester; n-Propyl gallate
C10H12O5
MFCD00002196
14,7859
n-Propyl 3,4,5-trihydroxybenzoate

Specifications

146° to 150°C
121-79-9
212.2
1877976
Gallic acid n-propyl ester; n-Propyl gallate
98%
2500g
C10H12O5
MFCD00002196
14,7859
n-Propyl 3,4,5-trihydroxybenzoate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302-H317
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P301+P312-P363-P321-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific treatment (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : LW8400000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit