Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Propyl Acetate, Reagent Grade, Reagents

Catalog No. 01338132
Click to view available options
Quantity:
4 L
10 L
20 L

N-Propyl Acetate, Reagent Grade, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 109-60-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
InChI Key YKYONYBAUNKHLG-UHFFFAOYSA-N
Synonym Propyl Ethanoate, Acetic Acid Propyl Ester
IUPAC Name propyl acetate
SMILES CCCOC(C)=O

Specifications

Melting Point -93°C
Color Colorless
Boiling Point 101.5°C
Packaging Steel Pail
Quantity 20 L
Specific Gravity 0.886
Grade Reagent
Physical Form Liquid
Chemical Name or Material N-Propyl Acetate, Reagent Grade
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.