Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
n-Propyl Alcohol, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.5%, Honeywell Burdick & Jackson
Click to view available options
Quantity:
19 L
204 L
Packaging:
Metal Can
Metal Drum
Specifications
CAS Max % | 100.0000% |
UN Number | UN1274 |
Beilstein | 1098242 |
Solubility Information | Miscible in Water at 20°C |
Formula Weight | 60.09 |
Percent Purity | >99.5% |
Grade | ACS Reagent |
Density | 0.8053 g/cm3 |
Boiling Point | 97.2°C |
Quantity | 19 L |
Show More |
Safety and Handling
ShelfLife | 547 days from date of manufacture |
DOTInformation | Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: n-Propanol |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction