Alfa Aesar™ n-Propyl decanoate, 98%

Manufacturer: Alfa Aesar™ B251590B

Quantity 1000g

 View more versions of this product

Catalog No. AAB251590B

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 30673-60-0
    Molecular Formula CH3(CH2)8CO2CH2CH2CH3
    Formula Weight 214.35
    MDL Number MFCD00056329
    Synonym Decanoic acid n-propyl ester; n-Propyl caprate
    Chemical Name or Material n-Propyl decanoate

    Specifications

    Formula Weight 214.35
    Flash Point 115°C (239°F)
    Quantity 1000g
    Molecular Formula CH3(CH2)8CO2CH2CH2CH3
    MDL Number MFCD00056329
    CAS 30673-60-0
    Synonym Decanoic acid n-propyl ester; n-Propyl caprate
    Boiling Point 254°C
    Chemical Name or Material n-Propyl decanoate
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit