Alfa Aesar™ n-Propyl hexanoate, 98%

Manufacturer: Alfa Aesar™ B2521522

Quantity 100g

 View more versions of this product

Catalog No. AAB2521522

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 626-77-7
    Molecular Formula C9H18O2
    Linear Formula CH3(CH2)4CO2CH2CH2CH3
    Formula Weight 158.24
    MDL Number MFCD00053803
    UN Number UN3272
    Synonym n-Propyl caproate; Hexanoic acid n-propyl ester
    Chemical Name or Material n-Propyl hexanoate

    Specifications

    Formula Weight 158.24
    TSCA TSCA
    Linear Formula CH3(CH2)4CO2CH2CH2CH3
    MDL Number MFCD00053803
    UN Number UN3272
    Health Hazard 1 Warning
    Flash Point 52°C (126°F)
    Boiling Point 187°C
    Molecular Formula C9H18O2
    CAS 626-77-7
    Synonym n-Propyl caproate; Hexanoic acid n-propyl ester
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
    Odor Fruit-like
    Melting Point −69°C
    Quantity 100g
    Chemical Name or Material n-Propyl hexanoate
    Assay 98%

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit