Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ n-Propyl hexanoate, 98%

Click to view available options
Quantity:
25g
100g
500g
626-77-7
C9H18O2
158.241
MFCD00053803
HTUIWRWYYVBCFT-UHFFFAOYSA-N
propyl caproate,caproic acid propyl ester,hexanoic acid, propyl ester,n-propyl hexanoate,propionyl hexanoate,n-propyl n-hexanoate,hexanoic acid propyl ester,natural,fema no. 2949,n-propyl hexanoaten
12293
CHEBI:87365
propyl hexanoate
CCCCCC(=O)OCCC
626-77-7
C9H18O2
158.241
MFCD00053803
HTUIWRWYYVBCFT-UHFFFAOYSA-N
propyl caproate,caproic acid propyl ester,hexanoic acid, propyl ester,n-propyl hexanoate,propionyl hexanoate,n-propyl n-hexanoate,hexanoic acid propyl ester,natural,fema no. 2949,n-propyl hexanoaten
12293
CHEBI:87365
propyl hexanoate
CCCCCC(=O)OCCC
Melting Point -69°C
Boiling Point 187°C
Quantity 500g
Odor Fruit-like
Flash Point 52°C (126°F)
Linear Formula CH3(CH2)4CO2CH2CH2CH3
UN Number UN3272
Formula Weight 158.24
Percent Purity 98%
Chemical Name or Material n-Propyl hexanoate

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

TSCA : TSCA