Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Propyl isocyanate, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
500g
Packaging:
Glass bottle
110-78-1
C4H7NO
85.11
MFCD00002045
OQURWGJAWSLGQG-UHFFFAOYSA-N
propyl isocyanate,propane, 1-isocyanato,n-propyl isocyanate,1-propyl isocyanate,propylisocyanate,isocyanic acid, propyl ester,m-propyl isocyanate,isocyanic acid propyl ester,1-propylisocyanate,1-isocyanato-propane
61033
1-isocyanatopropane
CCCN=C=O
110-78-1
C4H7NO
85.11
MFCD00002045
OQURWGJAWSLGQG-UHFFFAOYSA-N
propyl isocyanate,propane, 1-isocyanato,n-propyl isocyanate,1-propyl isocyanate,propylisocyanate,isocyanic acid, propyl ester,m-propyl isocyanate,isocyanic acid propyl ester,1-propylisocyanate,1-isocyanato-propane
61033
1-isocyanatopropane
CCCN=C=O
Quantity 25g
Density 0.8948g/mL
Packaging Glass bottle
Infrared Spectrum Authentic
Specific Gravity 0.8948
Flash Point −1°C
Color Yellow
Boiling Point 83.0°C to 84.0°C
Melting Point -30.0°C
Refractive Index 1.3940 to 1.3970
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH3CH2CH2NCO
Beilstein 04,I,366
Solubility Information Solubility in water: reacts vigorously
Formula Weight 85.11
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material n-Propyl isocyanate, 99%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

Recommended Storage : Product may develop some turbidity or precipitate