Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Propyltrichlorosilane, 98%, ACROS Organics™

Click to view available options
Quantity:
100mL
1L
Packaging:
Glass bottle
141-57-1
C3H7Cl3Si
177.53
DOEHJNBEOVLHGL-UHFFFAOYSA-N
propyltrichlorosilane,trichloro propyl silane,n-propyltrichlorosilane,silane, trichloropropyl,propyl trichlorosilane,trichloro-n-propylsilane,silane, propyltrichloro,unii-x16g85fi4y,hsdb 889,silane,trichloropropyl
8850
trichloro(propyl)silane
CCC[Si](Cl)(Cl)Cl
141-57-1
C3H7Cl3Si
177.53
DOEHJNBEOVLHGL-UHFFFAOYSA-N
propyltrichlorosilane,trichloro propyl silane,n-propyltrichlorosilane,silane, trichloropropyl,propyl trichlorosilane,trichloro-n-propylsilane,silane, propyltrichloro,unii-x16g85fi4y,hsdb 889,silane,trichloropropyl
8850
trichloro(propyl)silane
CCC[Si](Cl)(Cl)Cl
Infrared Spectrum Authentic
Packaging Glass bottle
Flash Point 31°C
Specific Gravity 1.19
Density 1.1900g/mL
Boiling Point 123.0°C to 124.0°C
Quantity 1L
Refractive Index 1.195
Color Colorless to Brown
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3CH2CH2SiCl3
Beilstein 04,36
Solubility Information Solubility in water: reacts
Formula Weight 177.53
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material n-Propyltrichlorosilane
Show More Show Less

GHS H Statement:
Flammable liquid and vapour.
Causes severe skin burns and eye damage.
Reacts violently with water.
Contact with water liberates toxic gas.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTRE or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger