Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Propyltrichlorosilane, 98%, ACROS Organics™

Click to view available options
Quantity:
100mL
1L
Packaging:
Glass bottle
141-57-1
C3H7Cl3Si
177.53
DOEHJNBEOVLHGL-UHFFFAOYSA-N
propyltrichlorosilane, trichloro propyl silane, n-propyltrichlorosilane, silane, trichloropropyl, propyl trichlorosilane, trichloro-n-propylsilane, silane, propyltrichloro, unii-x16g85fi4y, hsdb 889, silane,trichloropropyl
8850
trichloro(propyl)silane
CCC[Si](Cl)(Cl)Cl
141-57-1
C3H7Cl3Si
177.53
DOEHJNBEOVLHGL-UHFFFAOYSA-N
propyltrichlorosilane, trichloro propyl silane, n-propyltrichlorosilane, silane, trichloropropyl, propyl trichlorosilane, trichloro-n-propylsilane, silane, propyltrichloro, unii-x16g85fi4y, hsdb 889, silane,trichloropropyl
8850
trichloro(propyl)silane
CCC[Si](Cl)(Cl)Cl
Quantity 100mL
Specific Gravity 1.19
Packaging Glass bottle
Infrared Spectrum Authentic
Color Colorless to Brown
Density 1.1900g/mL
Flash Point 31°C
Refractive Index 1.195
Boiling Point 123.0°C to 124.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula CH3CH2CH2SiCl3
Beilstein 04,36
Solubility Information Solubility in water: reacts
Formula Weight 177.53
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material n-Propyltrichlorosilane
Show More Show Less

GHS H Statement
Flammable liquid and vapour.
Causes severe skin burns and eye damage.
Reacts violently with water.
Contact with water liberates toxic gas.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTRE or doctor/physician.
IF IN EYES: Rinse

GHS Signal Word: Danger

EINECSNumber : 205-489-6