Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Stearoyl-D-erythro-sphingosine, 99+%, synthetical, ACROS Organics™

Click to view available options
Quantity:
5mg
Packaging:
Glass bottle
2304-81-6
C36H71NO3
565.968
MFCD02259139
VODZWWMEJITOND-NXCSZAMKSA-N
n-2s,4e-1,3-dihydroxyoctadec-4-en-2-yl octadecanamide
5283565
CHEBI:72961
N-[(E,2S,3R)-1,3-dihydroxyoctadec-4-en-2-yl]octadecanamide
CCCCCCCCCCCCCCCCCC(=O)NC(CO)C(C=CCCCCCCCCCCCCC)O
2304-81-6
C36H71NO3
565.968
MFCD02259139
VODZWWMEJITOND-NXCSZAMKSA-N
n-2s,4e-1,3-dihydroxyoctadec-4-en-2-yl octadecanamide
5283565
CHEBI:72961
N-[(E,2S,3R)-1,3-dihydroxyoctadec-4-en-2-yl]octadecanamide
CCCCCCCCCCCCCCCCCC(=O)NC(CO)C(C=CCCCCCCCCCCCCC)O
Melting Point 99°C to 101°C
Quantity 5mg
Packaging Glass bottle
Infrared Spectrum Authentic
Color White
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Formula Weight 565.96
Physical Form Powder
Percent Purity ≥99.0%
Chemical Name or Material N-Stearoyl-D-erythro-sphingosine, 99+%
Show More Show Less