Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Succinylglycine, ≥95.0% (HPLC), MilliporeSigma™ Supelco™

$71.10 - $257.50

Specifications

Percent Purity ≥95.0% (HPLC)
Grade Analytical Standard
Linear Formula C6H9 NO5
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-4672
View Documents
MilliporeSigma Supelco
0188310MG
10 mg
Each for $71.10
Add to cart
 
11-101-4674
View Documents
MilliporeSigma Supelco
0188350MG
50 mg
Each for $146.00
Add to cart
 
11-101-4673
View Documents
MilliporeSigma Supelco
01883250MG
250 mg
Each for $257.50
Add to cart
 
Description

Description

Specifications

Specifications

≥95.0% (HPLC)
Analytical Standard
C6H9 NO5
Safety and Handling

Safety and Handling

ShelfLife : Limited shelf life, expiry date on the label

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit