Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-L-alanine N'-methoxy-N'-methylamide, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
87694-49-3
C10H20N2O4
232.28
PWQIGBOSLQHOBT-ZETCQYMHSA-N
boc-ala-nme ome,s-tert-butyl 1-methoxy methyl amino-1-oxopropan-2-yl carbamate,boc-ala-n och3 ch3,tert-butyl n-1s-1-methoxy methyl carbamoyl ethyl carbamate,s-2-boc-amino-n-methoxy-n-methylpropanamide,n-tert-butoxycarbonyl-l-alanine n'-methoxy-n'-methylamide,n2-tert-butoxycarbonyl-n-methoxy-n-methyl-l-alaninamide,2s-2-tert-butoxy carbonylamino-n-methoxy-n-methylpropanamide,s-tert-butyl 1-methoxy methyl amino-1-oxopropan-2-ylcarbamate,r-tert-butyl1-methoxy methyl amino-1-oxopropan-2-ylcarbamate
11806530
tert-butyl N-[(2S)-1-[methoxy(methyl)amino]-1-oxopropan-2-yl]carbamate
CC(C(=O)N(C)OC)NC(=O)OC(C)(C)C
87694-49-3
C10H20N2O4
232.28
PWQIGBOSLQHOBT-ZETCQYMHSA-N
boc-ala-nme ome,s-tert-butyl 1-methoxy methyl amino-1-oxopropan-2-yl carbamate,boc-ala-n och3 ch3,tert-butyl n-1s-1-methoxy methyl carbamoyl ethyl carbamate,s-2-boc-amino-n-methoxy-n-methylpropanamide,n-tert-butoxycarbonyl-l-alanine n'-methoxy-n'-methylamide,n2-tert-butoxycarbonyl-n-methoxy-n-methyl-l-alaninamide,2s-2-tert-butoxy carbonylamino-n-methoxy-n-methylpropanamide,s-tert-butyl 1-methoxy methyl amino-1-oxopropan-2-ylcarbamate,r-tert-butyl1-methoxy methyl amino-1-oxopropan-2-ylcarbamate
11806530
tert-butyl N-[(2S)-1-[methoxy(methyl)amino]-1-oxopropan-2-yl]carbamate
CC(C(=O)N(C)OC)NC(=O)OC(C)(C)C
Color White
Melting Point 143.5°C to 148.3°C
Specific Rotation -25.5
Quantity 5g
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 232.28
Physical Form Crystalline Powder or Powder
Percent Purity 98%
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-alanine N'-methoxy-N'-methylamide
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning