Alfa Aesar™ N-tert-Butyl-3,5-dimethylaniline, 97%

Manufacturer: Alfa Aesar™ H55164MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH55164MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 110993-40-3
    Formula Weight 177.29
    MDL Number MFCD07440135
    Chemical Name or Material N-tert-Butyl-3,5-dimethylaniline

    Specifications

    Formula Weight 177.29
    Boiling Point 69° to 70°C (0.75mmHg)
    Refractive Index 1.5190
    Density 0.901g/mL
    Physical Form Liquid
    MDL Number MFCD07440135
    CAS 110993-40-3
    Color Colorless to brown
    Quantity 250mg
    Chemical Name or Material N-tert-Butyl-3,5-dimethylaniline
    Assay 97%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit