Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-tert-Butyl-alpha-phenylnitrone, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
3376-24-7
C11H15NO
177.25
MFCD00008799
IYSYLWYGCWTJSG-FMIVXFBMSA-N
e-n-benzylidene-2-methylpropan-2-amine oxide
10313352
N-tert-butyl-1-phenylmethanimine oxide
CC(C)(C)[N+](=CC1=CC=CC=C1)[O-]
3376-24-7
C11H15NO
177.25
MFCD00008799
IYSYLWYGCWTJSG-FMIVXFBMSA-N
e-n-benzylidene-2-methylpropan-2-amine oxide
10313352
N-tert-butyl-1-phenylmethanimine oxide
CC(C)(C)[N+](=CC1=CC=CC=C1)[O-]
Packaging Glass bottle
Quantity 5g
Melting Point 71.0°C to 75.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Linear Formula C6H5CH=N(O)C(CH3)3
Merck Index 15,7162
Formula Weight 177.25
Percent Purity 98%
Chemical Name or Material N-tert-Butyl-α-phenylnitrone
Show More Show Less

EINECSNumber : 222-168-6