Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ n-Tetradecanamide, 98%

Click to view available options
Quantity:
5g
25g
100g
638-58-4
C14H29NO
227.392
MFCD00025533
QEALYLRSRQDCRA-UHFFFAOYSA-N
myristamide,n-tetradecanamide,myristic amide,myristic acid amide,tetradecylamide,unii-a269j8qg0o,myristicamide,tetradecanoic acid amide,chembl88158
69492
tetradecanamide
CCCCCCCCCCCCCC(=O)N
638-58-4
C14H29NO
227.392
MFCD00025533
QEALYLRSRQDCRA-UHFFFAOYSA-N
myristamide,n-tetradecanamide,myristic amide,myristic acid amide,tetradecylamide,unii-a269j8qg0o,myristicamide,tetradecanoic acid amide,chembl88158
69492
tetradecanamide
CCCCCCCCCCCCCC(=O)N
Quantity 100g
Melting Point 105°C to 107°C
Formula Weight 227.39
Percent Purity 98%
Chemical Name or Material n-Tetradecanamide