Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Tosylpyrrolidone, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
10019-95-1
C11H13NO3S
239.289
MFCD00010421
AAXCXJDQWSSCHL-UHFFFAOYSA-N
1-p-toluenesulfonyl-2-pyrrolidinone,1-tosylpyrrolidin-2-one,n-tosylpyrrolidone,1-4-methylphenyl sulfonyl pyrrolidin-2-one,1-4-methylbenzenesulfonyl pyrrolidin-2-one,1-tosylpyrrolidine-2-one,n-p-tosyl-pyrrolidin-2-one,1-4-methylphenyl sulfonylpyrrolidin-2-one,1-4-methylphenyl sulfonyl-2-pyrrolidinone,2-pyrrolidinone,1-4-methylphenyl sulfonyl
563071
1-(4-methylphenyl)sulfonylpyrrolidin-2-one
CC1=CC=C(C=C1)S(=O)(=O)N2CCCC2=O
10019-95-1
C11H13NO3S
239.289
MFCD00010421
AAXCXJDQWSSCHL-UHFFFAOYSA-N
1-p-toluenesulfonyl-2-pyrrolidinone,1-tosylpyrrolidin-2-one,n-tosylpyrrolidone,1-4-methylphenyl sulfonyl pyrrolidin-2-one,1-4-methylbenzenesulfonyl pyrrolidin-2-one,1-tosylpyrrolidine-2-one,n-p-tosyl-pyrrolidin-2-one,1-4-methylphenyl sulfonylpyrrolidin-2-one,1-4-methylphenyl sulfonyl-2-pyrrolidinone,2-pyrrolidinone,1-4-methylphenyl sulfonyl
563071
1-(4-methylphenyl)sulfonylpyrrolidin-2-one
CC1=CC=C(C=C1)S(=O)(=O)N2CCCC2=O
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 1g
Color White to Yellow
Formula Weight 239.29
Physical Form Crystalline Powder
Chemical Name or Material N-Tosylpyrrolidone