Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Tridecane, 99+%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
500mL
Packaging:
Glass bottle
629-50-5
C13H28
184.36
MFCD00008979
IIYFAKIEWZDVMP-UHFFFAOYSA-N
n-tridecane,tridekan,alkanes, c12-14,dodecane, methyl,unii-a3lzf0l939,tridecane, n,trd,tridecane, analytical standard,tridecan,tridecyl group
12388
CHEBI:35998
tridecane
CCCCCCCCCCCCC
629-50-5
C13H28
184.36
MFCD00008979
IIYFAKIEWZDVMP-UHFFFAOYSA-N
n-tridecane,tridekan,alkanes, c12-14,dodecane, methyl,unii-a3lzf0l939,tridecane, n,trd,tridecane, analytical standard,tridecan,tridecyl group
12388
CHEBI:35998
tridecane
CCCCCCCCCCCCC
Flash Point 79°C
Color Colorless
Quantity 25mL
Melting Point -5°C to 4°C
Boiling Point 234.0°C
Packaging Glass bottle
Density 0.7500g/mL
Refractive Index 1.4240 to 1.4260
Infrared Spectrum Authentic
Specific Gravity 0.75
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99% min. (GC)
Linear Formula CH3(CH2)11CH3
Beilstein 01, 171
Solubility Information Solubility in water: negligible. Other solubilities: soluble in alcohol
Formula Weight 184.36
Physical Form Liquid
Percent Purity 99+%
Chemical Name or Material n-Tridecane
Show More Show Less

GHS H Statement
May be fatal if swallowed and enters airways.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.

GHS Signal Word: Danger

EINECSNumber : 211-093-4