Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Tridecane, 99+%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
500mL
Packaging:
Glass bottle
629-50-5
C13H28
184.36
MFCD00008979
IIYFAKIEWZDVMP-UHFFFAOYSA-N
n-tridecane,tridekan,alkanes, c12-14,dodecane, methyl,unii-a3lzf0l939,tridecane, n,trd,tridecane, analytical standard,tridecan,tridecyl group
12388
CHEBI:35998
tridecane
CCCCCCCCCCCCC
629-50-5
C13H28
184.36
MFCD00008979
IIYFAKIEWZDVMP-UHFFFAOYSA-N
n-tridecane,tridekan,alkanes, c12-14,dodecane, methyl,unii-a3lzf0l939,tridecane, n,trd,tridecane, analytical standard,tridecan,tridecyl group
12388
CHEBI:35998
tridecane
CCCCCCCCCCCCC
Melting Point -5°C to 4°C
Packaging Glass bottle
Flash Point 79°C
Density 0.7500g/mL
Quantity 500mL
Infrared Spectrum Authentic
Boiling Point 234.0°C
Specific Gravity 0.75
Refractive Index 1.4240 to 1.4260
Color Colorless
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula CH3(CH2)11CH3
Beilstein 01, 171
Solubility Information Solubility in water: negligible. Other solubilities: soluble in alcohol
Formula Weight 184.36
Physical Form Liquid
Percent Purity ≥99%
Chemical Name or Material n-Tridecane
Show More Show Less

GHS H Statement:
May be fatal if swallowed and enters airways.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.