Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
n-Tridecane, EMD-Millipore
Click to view available options
Quantity:
5mL
Packaging:
Glass ampoule
Specifications
Assay Percent Range | ≥99.5% |
Linear Formula | CH3(CH2)11CH3 |
Formula Weight | 184.36 |
Explosion Limit | 0.6 to 6.5 %(V) |
Ignition Point | >204°C |
Packaging | Glass ampoule |
Vapor Pressure | < - 0.5 hPa (20°C) |
Viscosity | 4.28mm2/sec., kinematic (20°C) |
Density | Density: 0.757g/cm3 (20°) |
Quantity | 5mL |
Show More |
Safety and Handling
Recommended Storage : Store without restrictions