Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Tridecanoic Acid 97%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
50g
Packaging:
Glass bottle
638-53-9
C13H26O2
214.35
MFCD00002741
SZHOJFHSIKHZHA-UHFFFAOYSA-N
n-tridecanoic acid,tridecylic acid,n-tridecoic acid,unii-19936liy2v,anion,tridecanoic acid,wln: qv12,c10-16 fatty acids,fatty acids, c10-16,carboxylic acids, c10-16
12530
CHEBI:45919
tridecanoic acid
CCCCCCCCCCCCC(=O)O
638-53-9
C13H26O2
214.35
MFCD00002741
SZHOJFHSIKHZHA-UHFFFAOYSA-N
n-tridecanoic acid,tridecylic acid,n-tridecoic acid,unii-19936liy2v,anion,tridecanoic acid,wln: qv12,c10-16 fatty acids,fatty acids, c10-16,carboxylic acids, c10-16
12530
CHEBI:45919
tridecanoic acid
CCCCCCCCCCCCC(=O)O
Quantity 25g
Packaging Glass bottle
Boiling Point 236°C (100.0mmHg)
Flash Point 113°C
Color White to Yellow
Melting Point 40°C to 45°C
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3(CH2)11CO2H
Beilstein 02, 364
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 214.35
Physical Form Crystalline Chunks, Flakes or Powder
Percent Purity ≥96.0%
Chemical Name or Material n-Tridecanoic acid, 97%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 211-341-1

RTECSNumber : YD3850000

TSCA : TSCA