Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Trifluoroacetyl-D-glucosamine 96.0+%, TCI America™

Click to view available options
Quantity:
1g
5g
36875-26-0
C8H12F3NO6
MFCD00059807
2-Deoxy-2-trifluoroacetamido-D-glucopyranose
36875-26-0
C8H12F3NO6
MFCD00059807
2-Deoxy-2-trifluoroacetamido-D-glucopyranose
Quantity 1g
Color Undesignated
Formula Weight 275.18
Physical Form Crystal-Powder at 20°C
Percent Purity ≥96.0% (N)
Chemical Name or Material N-Trifluoroacetyl-D-glucosamine