Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(Trimethylsilyl)acetamide, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
13435-12-6
C5H13NOSi
131.25
MFCD00008671
LWFWUJCJKPUZLV-UHFFFAOYSA-N
n-trimethylsilyl acetamide,trimethylsilylacetamide,acetamide, n-trimethylsilyl,tms acetamide,acetylamino trimethylsilane,trimethylsilyl acetamide,acetamide, tms derivative,n-trimethylsilyl-acetamide,n-tms-acetamide,n-trimethylsilylacetoamide
25989
N-trimethylsilylacetamide
CC(=O)N[Si](C)(C)C
13435-12-6
C5H13NOSi
131.25
MFCD00008671
LWFWUJCJKPUZLV-UHFFFAOYSA-N
n-trimethylsilyl acetamide,trimethylsilylacetamide,acetamide, n-trimethylsilyl,tms acetamide,acetylamino trimethylsilane,trimethylsilyl acetamide,acetamide, tms derivative,n-trimethylsilyl-acetamide,n-tms-acetamide,n-trimethylsilylacetoamide
25989
N-trimethylsilylacetamide
CC(=O)N[Si](C)(C)C
Color White to Yellow
Boiling Point 84°C (18.0mmHg)
Flash Point 70°C
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 25g
Melting Point 40°C to 46°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3CONHSi(CH3)3
Fieser 01,1235; 16,178
Solubility Information Solubility in water: reacts
Formula Weight 131.25
Physical Form Crystalline Mass
Percent Purity 97%
Chemical Name or Material N-(Trimethylsilyl)acetamide
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable solid.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 236-565-7