Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[(Trimethylsilyl)methyl]benzylamine, 95%, Alfa Aesar™

Click to view available options
Quantity:
5g
25g
100g
53215-95-5
C11H19NSi
193.365
MFCD00012192
WECLUYCAWLJMKM-UHFFFAOYSA-N
n-trimethylsilylmethyl benzylamine,n-trimethylsilyl methyl benzylamine,benzyl trimethylsilyl methyl amine,n-benzyl-1-trimethylsilyl methylamine,1-phenyl-n-trimethylsilylmethyl methanamine,benzyl trimethylsilylmethyl amine,n-benzyl-1-trimethylsilyl methanamine,benzenemethanamine, n-trimethylsilyl methyl,n-benzylaminomethyltrimethylsilane,n-benzyl-n-trimethylsilyl methyl amine
353458
1-phenyl-N-(trimethylsilylmethyl)methanamine
C[Si](C)(C)CNCC1=CC=CC=C1
53215-95-5
C11H19NSi
193.365
MFCD00012192
WECLUYCAWLJMKM-UHFFFAOYSA-N
n-trimethylsilylmethyl benzylamine,n-trimethylsilyl methyl benzylamine,benzyl trimethylsilyl methyl amine,n-benzyl-1-trimethylsilyl methylamine,1-phenyl-n-trimethylsilylmethyl methanamine,benzyl trimethylsilylmethyl amine,n-benzyl-1-trimethylsilyl methanamine,benzenemethanamine, n-trimethylsilyl methyl,n-benzylaminomethyltrimethylsilane,n-benzyl-n-trimethylsilyl methyl amine
353458
1-phenyl-N-(trimethylsilylmethyl)methanamine
C[Si](C)(C)CNCC1=CC=CC=C1
Quantity 25g
Refractive Index 1.4960
Density 0.88g/mL
Color Yellow
Boiling Point 89°C to 90°C (5mmHg)
Beilstein 2802488
Formula Weight 193.36
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material N-[(Trimethylsilyl)methyl]benzylamine