Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[Tris(hydroxymethyl)methyl]acrylamide, 99%, Alfa Aesar™

Click to view available options
Quantity:
5g
25g
100g
13880-05-2
C7H13NO4
175.184
MFCD00081045
MVBJSQCJPSRKSW-UHFFFAOYSA-N
n-tris hydroxymethyl methyl acrylamide,n-1,3-dihydroxy-2-hydroxymethyl propan-2-yl acrylamide,tris-acrylamide,n-acryloyltris hydroxymethyl aminomethane,n-acryloyl-tris hydroxymethyl aminomethane,n-1,3-dihydroxy-2-hydroxymethyl propan-2-yl prop-2-enamide,zlchem 121,acryloylaminomethylidynetrimethanol,n-1,3-dihydroxy-2-hydroxymethyl-2-propanyl acrylamide
83788
N-[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]prop-2-enamide
C=CC(=O)NC(CO)(CO)CO
13880-05-2
C7H13NO4
175.184
MFCD00081045
MVBJSQCJPSRKSW-UHFFFAOYSA-N
n-tris hydroxymethyl methyl acrylamide,n-1,3-dihydroxy-2-hydroxymethyl propan-2-yl acrylamide,tris-acrylamide,n-acryloyltris hydroxymethyl aminomethane,n-acryloyl-tris hydroxymethyl aminomethane,n-1,3-dihydroxy-2-hydroxymethyl propan-2-yl prop-2-enamide,zlchem 121,acryloylaminomethylidynetrimethanol,n-1,3-dihydroxy-2-hydroxymethyl-2-propanyl acrylamide
83788
N-[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]prop-2-enamide
C=CC(=O)NC(CO)(CO)CO
Applications
N-[Tris(hydroxymethyl)methyl] acrylamide is used to prepare N-(5-hydroxymethyl-2-phenyl-[1,3]dioxan-5-yl)-acrylamide by reaction with benzaldehyde.

Solubility
Soluble in water.

Notes
Hygroscopic. Light sensitive. Incompatible with strong oxdizing agents.
Melting Point 134°C to 138°C
Sensitivity Hygroscopic; Light sensitive
Quantity 5g
Linear Formula CH2=CHCONHC(CH2OH)3
Solubility Information Soluble in water.
Formula Weight 175.18
Percent Purity 99%
Chemical Name or Material N-[Tris(hydroxymethyl)methyl]acrylamide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 237-645-4

TSCA : No

Recommended Storage : Keep cold