Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Vinyl-2-pyrrolidinone, Stabilized 98%, ACROS Organics™

Click to view available options
Quantity:
5mL
500mL
2.5L
10L
88-12-0
C6H9NO
111.144
MFCD00003197
WHNWPMSKXPGLAX-UHFFFAOYSA-N
n-vinyl-2-pyrrolidone,n-vinylpyrrolidone,1-vinyl-2-pyrrolidone,1-vinylpyrrolidin-2-one,n-vinyl-2-pyrrolidinone,vinylpyrrolidone,2-pyrrolidinone, 1-ethenyl,1-vinyl-2-pyrrolidinone,n-vinylpyrrolidinone,1-vinylpyrrolidone
6917
CHEBI:82551
1-ethenylpyrrolidin-2-one
C=CN1CCCC1=O
88-12-0
C6H9NO
111.144
MFCD00003197
WHNWPMSKXPGLAX-UHFFFAOYSA-N
n-vinyl-2-pyrrolidone,n-vinylpyrrolidone,1-vinyl-2-pyrrolidone,1-vinylpyrrolidin-2-one,n-vinyl-2-pyrrolidinone,vinylpyrrolidone,2-pyrrolidinone, 1-ethenyl,1-vinyl-2-pyrrolidinone,n-vinylpyrrolidinone,1-vinylpyrrolidone
6917
CHEBI:82551
1-ethenylpyrrolidin-2-one
C=CN1CCCC1=O
Color Colorless to Yellow
Infrared Spectrum Authentic
Quantity 10L
Water 0.2% max. (K.F.)
Density 1.0430g/mL
Stabilizer 0.1% flaked caustic soda
Refractive Index 1.5110 to 1.5140 (20°C, 589nm)
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 111.14
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material N-Vinyl-2-pyrrolidone, 98%
Show More Show Less

GHS H Statement:
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye damage.
Toxic if inhaled.
Toxic in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 201-800-4

RTECSNumber : UY6107000

TSCA : TSCA

Recommended Storage : Product may darken on storage