Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Vinylphthalimide, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
3485-84-5
C10H7NO2
173.171
MFCD00078446
IGDLZDCWMRPMGL-UHFFFAOYSA-N
n-vinylphthalimide,2-vinylisoindoline-1,3-dione,phthalimide, n-vinyl,1h-isoindole-1,3 2h-dione, 2-ethenyl,2-vinyl-1h-isoindole-1,3 2h-dione,2-ethenyl-1h-isoindole-1,3 2h-dione,2-ethenyl-1,3-dihydro-1,3-dioxoisoindole,2-ethenyl-2,3-dihydro-1h-isoindole-1,3-dione,2-vinylbenzo c azolidine-1,3-dione,n-vinylphtalimide
77035
2-ethenylisoindole-1,3-dione
C=CN1C(=O)C2=CC=CC=C2C1=O
3485-84-5
C10H7NO2
173.171
MFCD00078446
IGDLZDCWMRPMGL-UHFFFAOYSA-N
n-vinylphthalimide,2-vinylisoindoline-1,3-dione,phthalimide, n-vinyl,1h-isoindole-1,3 2h-dione, 2-ethenyl,2-vinyl-1h-isoindole-1,3 2h-dione,2-ethenyl-1h-isoindole-1,3 2h-dione,2-ethenyl-1,3-dihydro-1,3-dioxoisoindole,2-ethenyl-2,3-dihydro-1h-isoindole-1,3-dione,2-vinylbenzo c azolidine-1,3-dione,n-vinylphtalimide
77035
2-ethenylisoindole-1,3-dione
C=CN1C(=O)C2=CC=CC=C2C1=O
Quantity 1g
Packaging Glass bottle
Melting Point 84°C to 88°C
Infrared Spectrum Authentic
Color White to Yellow
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 173.17
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material N-Vinylphthalimide
Show More Show Less

GHS P Statement May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection.

Warning