Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Z-Ethanolamine, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
77987-49-6
C10H13NO3
195.22
MFCD00191060
SAGINAGERRNGGV-UHFFFAOYSA-N
benzyl n-2-hydroxyethyl carbamate,z-glycinol,benzyl 2-hydroxyethyl carbamate,n-z-ethanolamine,2-cbz-amino-1-ethanol,benzyl 2-hydroxyethylcarbamate,2-z-amino ethanol,n-2-hydroxyethyl phenylmethoxy carboxamide,n-2-hydroxyethyl carbamic acid benzyl ester
280458
benzyl N-(2-hydroxyethyl)carbamate
C1=CC=C(C=C1)COC(=O)NCCO
77987-49-6
C10H13NO3
195.22
MFCD00191060
SAGINAGERRNGGV-UHFFFAOYSA-N
benzyl n-2-hydroxyethyl carbamate,z-glycinol,benzyl 2-hydroxyethyl carbamate,n-z-ethanolamine,2-cbz-amino-1-ethanol,benzyl 2-hydroxyethylcarbamate,2-z-amino ethanol,n-2-hydroxyethyl phenylmethoxy carboxamide,n-2-hydroxyethyl carbamic acid benzyl ester
280458
benzyl N-(2-hydroxyethyl)carbamate
C1=CC=C(C=C1)COC(=O)NCCO
Boiling Point 170°C (0.5 Torr)
Melting Point 61°C to 64°C
Quantity 25g
Infrared Spectrum Authentic
Color White
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 195.22
Physical Form Powder
Percent Purity 98%
Chemical Name or Material N-Z-Ethanolamine
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning