Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Z-Ethylenediamine hydrochloride, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
18807-71-1
C10H14N2O2·ClH
230.69
QMLKQXIAPAAIEJ-UHFFFAOYSA-N
benzyl n-2-aminoethyl carbamate hydrochloride,n-cbz-ethylenediamine hydrochloride,benzyl 2-aminoethyl carbamate hydrochloride,n-carbobenzoxy-1,2-diaminoethane hydrochloride,n-z-ethylenediamine hydrochloride,n-benzyloxycarbonyl-1,2-diaminoethane hydrochloride,n-z-1,2-diaminoethane hydrochloride,n-cbz-1,2-diaminoethane hydrochloride,benzyl 2-aminoethyl carbamate hcl,n-benzyloxycarbonylethylenediamine hydrochloride
12715871
benzyl N-(2-aminoethyl)carbamate;hydrochloride
C1=CC=C(C=C1)COC(=O)NCCN.Cl
18807-71-1
C10H14N2O2·ClH
230.69
QMLKQXIAPAAIEJ-UHFFFAOYSA-N
benzyl n-2-aminoethyl carbamate hydrochloride,n-cbz-ethylenediamine hydrochloride,benzyl 2-aminoethyl carbamate hydrochloride,n-carbobenzoxy-1,2-diaminoethane hydrochloride,n-z-ethylenediamine hydrochloride,n-benzyloxycarbonyl-1,2-diaminoethane hydrochloride,n-z-1,2-diaminoethane hydrochloride,n-cbz-1,2-diaminoethane hydrochloride,benzyl 2-aminoethyl carbamate hcl,n-benzyloxycarbonylethylenediamine hydrochloride
12715871
benzyl N-(2-aminoethyl)carbamate;hydrochloride
C1=CC=C(C=C1)COC(=O)NCCN.Cl
Melting Point 170°C to 173°C
Color Brown to Yellow
Quantity 1g
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (HPLC)
Formula Weight 230.69
Physical Form Solution Product May Develop Some Turbidity or Precipitate
Percent Purity 95%
Chemical Name or Material N-Z-Ethylenediamine hydrochloride
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.

GHS Signal Word: Warning