Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ Normorphine Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: MilliporeSigma Supelco N0061ML
Description
Normorphine is a primary urinary metabolite of the opiate analgesic morphine. This Snap-N-Spike™ reference solution is applicable for use in LC/MS or GC/MS applications such as pain prescription monitoring, urine drug testing, clinical toxicology, or forensic analysis. Morphine, used to relieve both acute and chronic severe pain is known under trade names MS Contin™, Kadian™, Oramorph™, and Roxanol.Specifications
67-56-1, 466-97-7 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
271.31 | |
1 mL |
C16H17NO3 | |
1.0 mg/mL in methanol | |
Ampule | |
9.7°C (49.5°F) - closed cup |
Safety and Handling
H331-H370
H311
Recommended Storage : 2 to 8°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation
Spot an opportunity for improvement?
Provide Content Correction