Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ Nandrolone Solution, 1.0 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: MilliporeSigma Supelco N0501ML
Description
An analytical reference standard applicable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from sports testing and urine drug testing to forensic analysis. Nandrolone is an anabolic steroid sold commercially as Deca-Durabolin™. This performance enhancing drug is disallowed in sports competition.Specifications
75-05-8, 434-22-0 | |
MFCD00003664 | |
1.0 mg/mL in acetonitrile | |
Ampule | |
2°C (35.6°F) - closed cup |
C18H26O2 | |
UN 1648 3 / PGII | |
274.40 | |
1 mL |
Safety and Handling
H332-H319
H312
Recommended Storage : -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation
Spot an opportunity for improvement?
Provide Content Correction