Learn More
CERILLIANT™ (R,S)-Norcotinine-13C3 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco N0951ML
Catalog No. 01-001-756
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Norcotinine, or desmethylcotinine, is a urinary metabolite of nornicotine and cotinine found in smokers and users of other nicotine products including chewing tobacco, snuff and electronic cigarettes. This internal standard is suitable for quantitation of norcotinine levels in urine, serum, saliva or plasma by GC/MS or LC/MS for urine drug testing, clinical toxicology, nicotine testing or forensic analysis.
Specifications
Specifications
67-56-1,1020719-70-3
9.7°C (49.5°F) - closed cup
13C3C6H10N2O
1 mL
UN 1230 - class 3 - PG 2 - Methanol
100 μg/mL in methanol
165.17
Ampule
-20°C
Safety and Handling
Safety and Handling
H331-H370
H311
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation