Learn More
N1-Phosphohistidine (1-pHis) Rabbit anti-Human, KHL, Clone: SC50-3, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma ZRB13414X25UL
 View more versions of this product
Catalog No. ZRB13414XMI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
N1-Phosphohistidine (1-pHis) Monoclonal antibody specifically detects N1-Phosphohistidine (1-pHis) in Human samples. It is validated for Western Blotting.
Specifications
Specifications
N1-Phosphohistidine (1-pHis)
SC50-3
KHL
Phosphohistidine
KLH-conjugated library of random peptides containing non-hydrolyzable phosphohistidine analogue 1-pTza.
∼45 kDa observed; may vary depending on the histidine-phosphorylated proteins. Uncharacterized bands may be observed in some lysate(s).
4 x 25 μL
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Monoclonal
Human
Western Blot
300 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Purified
Rabbit
IgG
Protein A Purified
Signaling
Primary
4X