Learn More
CERILLIANT™ Nortetrazepam-13C2,15N Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco N1231ML
Catalog No. 01-001-775
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
A stable-labeled internal standard suitable for quantitation of nortetrazepam levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, forensic analysis, or isotope dilution methods. Nortetrazepam is a benzodiazepine derivative and one of the major metabolites of tetrazepam.
Specifications
Specifications
67-56-1
277.72
Ampule
-20°C
100 μg/mL in methanol
C1313C2H15ClN15NO
1 mL
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation