Learn More
CERILLIANT™ Norcarfentanil-13C6 oxalate Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco N1291EA
Description
This stable labeled internal standard is suitable for quantitation of Norcarfentanil in fentanyl GC/MS or LC/MS methods for applications in clinical toxicology, forensic analysis, prescription monitoring or urine drug testing. Norcarfentanil is a major urinary metabolite of carfentanil, a potent synthetic opioid analgesic drug structurally related to fentanyl. It is used in combination with other opioids and analgesics to induce or supplement general anesthesia.Specifications
67-56-1 | |
386.35 | |
Ampule | |
-20°C |
100 μg/mL in methanol | |
13C6C10H22N2O3 · C2H2O4 | |
0.5 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation