Learn More
CERILLIANT™ Norfentanyl-13C6 oxalate Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco N1300.5ML
Description
An internal standard suitable for norfentanyl testing of urine, blood, or hair samples or isotope dilution methods by GC/MS or LC/MS in applications such as pain prescription monitoring, clinical toxicology, urine drug testing, or forensic analysis. Norfentanyl is a major urinary metabolite of fentanyl, a potent synthetic analgesic sold under various trade names including Sublimaze, Duragesic, Actiq, and Fentora.Specifications
67-56-1 | |
328.31 | |
Ampule | |
-20°C |
100 μg/mL in methanol (as free base) | |
C813C6H20N2O · C2H2O4 | |
0.5 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation