Learn More
CERILLIANT™ Nimetazepam-D3 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco N1311ML
Catalog No. 01-001-780
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
A stable-labeled internal standard suitable for quantitation of Nimetazepam levels in urine, serum, or plasma by LC/MS or GC/MS for forensic analysis, clinical toxicology, or urine drug testing. Nimetazepam, marketed as the benzodiazepine drug Erimin, is prescribed for the treatment of stress or short-term severe insomnia in patients who have difficulty falling asleep or maintaining sleep. Although not used therapeutically in the US, the drug is known for its use to soften the comedown after use of methamphetamine or other stimulants.
Specifications
Specifications
67-56-1
9.7°C (49.5°F) - closed cup
C16H10D3N3O3
1 mL
100 μg/mL in methanol
298.31
Ampule
-20°C
Safety and Handling
Safety and Handling
H331-H370
H311
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation