Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris™ N20C hydrochloride

Non-competitive NMDA receptor open-channel blocker

Manufacturer:  Tocris™ 2213/50

 View more versions of this product

Catalog No. 22-135-0


  Check Availability
Add to cart

Disclaimers:

For Research Use Only


Specifications

Description

Selective, non-competitive NMDA receptor antagonist (IC50 = 5µM); binds to the receptor-associated ion channel and prevents glutamate-induced Ca2+ influx. Displays neuroprotective activity in vivo. Brain-penetrant.
  • Chemical Name: 2-[(3,3-Diphenylpropyl)amino]acetamide hydrochloride
  • Formula: C17H20N2O.HCl
  • CAS Number: 1177583-87-7
  • Solubility: Soluble to 100mM in water
  • Purity: >98
Description & Specifications

Specifications

N20C hydrochloride
>98
50mg
N20C hydrochloride
304.82
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit