Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N3-Phosphohistidine (3-pHis) Rabbit anti-Human, KHL, Clone: SC39-6, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen N3-Phosphohistidine (3-pHis)
Applications Western Blot
Conjugate KHL
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB135125MI
View Documents
MilliporeSigma
ZRB135125UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB13514XMI
View Documents
MilliporeSigma
ZRB13514X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

N3-Phosphohistidine (3-pHis) Monoclonal antibody specifically detects N3-Phosphohistidine (3-pHis) in Human samples. It is validated for Western Blotting.
Specifications

Specifications

N3-Phosphohistidine (3-pHis)
KHL
Rabbit
Phosphohistidine
Monoclonal
Protein A Purified
Western Blot
Purified
Human
KLH-conjugated library of random peptides containing non-hydrolyzable phosphohistidine analogue 1-pTza.
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit