Learn More
Valiant™ N3200 Low Dermatitis Blue Nitrile Gloves
Patented low derma technology eliminates chemical accelerators that are commonly found in nitrile, reducing the risk of Type 1 & Type 4 hypersensitivities.
$31.64 - $316.40
Specifications
Accelerator Free Yes
Chemotherapeutic Rated Yes
Color Blue
Disposable Yes
Exterior Finish Partially Textured
View More Specs
Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-444-030
View Documents
Valiant™
N3200LDS
Small
Case for $316.40
Add to cart
19-444-031
View Documents
Valiant™
N3200LDM
Medium
Case for $316.40
Add to cart
19-444-032
View Documents
Valiant™
N3200LDL
Large
Case for $316.40
Add to cart
19-444-033
View Documents
Valiant™
N3200LDXL
X-Large
Pack of 180 for $31.64
Case of 10 PK for $316.40
Add to cart
Description
Description
  • Exam grade nitrile gloves
  • Low dermatitis potential
  • Textured fingertips
  • Chemotherapy tested, results located on back of the box
  • Tested for resistance to Fentanyl Citrate
  • Accelerator-free
Specifications
Specifications
Yes
Blue
Partially Textured
Yes
9.5 in.
Powder Free
Non-sterile
Yes
Yes
3 mil
Yes
Nitrile
N3200