Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N4-Acetylcytosine, 98%, Acros Organics™

Click to view available options
Quantity:
5g
25g
14631-20-0
C6H7N3O2
153.14
IJCKBIINTQEGLY-UHFFFAOYSA-N
n4-acetylcytosine,n-acetylcytosine,n-2-oxo-1,2-dihydropyrimidin-4-yl acetamide,acetamide, n-1,2-dihydro-2-oxo-4-pyrimidinyl,n 4-acetylcytosine,n-2-oxo-1h-pyrimidin-6-yl acetamide,n-2-hydroxypyrimidin-4-yl acetamide,n-2-oxo-3h-pyrimidin-4-yl acetamide,4-acetylamino-1,2-dihydro-2-pyrimidone
99309
N-(2-oxo-1H-pyrimidin-6-yl)acetamide
CC(=O)NC1=CC=NC(=O)N1
14631-20-0
C6H7N3O2
153.14
IJCKBIINTQEGLY-UHFFFAOYSA-N
n4-acetylcytosine,n-acetylcytosine,n-2-oxo-1,2-dihydropyrimidin-4-yl acetamide,acetamide, n-1,2-dihydro-2-oxo-4-pyrimidinyl,n 4-acetylcytosine,n-2-oxo-1h-pyrimidin-6-yl acetamide,n-2-hydroxypyrimidin-4-yl acetamide,n-2-oxo-3h-pyrimidin-4-yl acetamide,4-acetylamino-1,2-dihydro-2-pyrimidone
99309
N-(2-oxo-1H-pyrimidin-6-yl)acetamide
CC(=O)NC1=CC=NC(=O)N1
Quantity 5g
Melting Point >300.0°C
Infrared Spectrum Authentic
Water 1% max.
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 153.14
Percent Purity 98%
Chemical Name or Material N4-Acetylcytosine

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning