Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ N6AMT1 Monoclonal Antibody (1C4)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA521764
Description
Peptide Sequence: LETARCNKVH IQPVITDLVG SHGIEAAWAG GRNGREVMDR FFPLVPDLLS PRGLFYLVTI KENNPEEILK IMKTKGLQGT TALSRQAGQE TLSVLKFTKS.
Heterodimeric methyltransferase that catalyzes N5-methylation of ETF1 on 'Gln-185', using S-adenosyl L-methionine as methyl donor. ETF1 needs to be complexed to ERF3 in its GTP-bound form to be efficiently methylated. May play a role in the modulation of arsenic-induced toxicity. May be involved in the conversion of monomethylarsonous acid (3+) into the less toxic dimethylarsonic acid.
Specifications
N6AMT1 | |
Monoclonal | |
0.2-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q9Y5N5 | |
N6AMT1 | |
N6AMT1 (NP_877426, 87 a.a. ~ 186 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Western Blot | |
1C4 | |
Unconjugated | |
N6AMT1 | |
5830445C04Rik; C21orf127; HemK methyltransferase family member 2; HEMK2; m.HsaHemK2P; Methylarsonite methyltransferase N6AMT1; Methyltransferase N6AMT1; methyltransferase N6AMT1; hemK methyltransferase family member 2; M-N6DNA MTase A; MTQ2; N(6)-adenine-specific DNA methyltransferase 1; N(6)-adenine-specific DNA methyltransferase 1-like; N-6 adenine-specific DNA methyltransferase 1; N-6 adenine-specific DNA methyltransferase 1 (putative); N6AMT; N6amt1; N6-DNA methyltransferase A; N6-DNA-methyltransferase; PRED28; Protein N(5)-glutamine methyltransferase; RGD1311843 | |
Mouse | |
Affinity chromatography | |
RUO | |
29104 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction