Learn More
Invitrogen™ N6AMT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121076
Description
Positive test controls include: Rat kidney. Immunogen sequence: MAGENFATPF HGHVGRGAFS DVYEPAEDTF LLLNALEAAA AELAGVEICL EVGSGSGVVS AFLASMIGPQ ALYMCTDINP EAAACTLETA RCNKVHIQPV ITDLVGSHGI EAAWAGGRNG REVMDRFFPL VPDLLSPRGL FYLVTIKENN PEEILKIMKT KGLQGTTALS RQAGQETLSV LKFTKS.
Heterodimeric methyltransferase that catalyzes N5-methylation of ETF1 on 'Gln-185', using S-adenosyl L-methionine as methyl donor. ETF1 needs to be complexed to ERF3 in its GTP-bound form to be efficiently methylated. May play a role in the modulation of arsenic-induced toxicity. May be involved in the conversion of monomethylarsonous acid (3+) into the less toxic dimethylarsonic acid.
Specifications
N6AMT1 | |
Polyclonal | |
Unconjugated | |
N6AMT1 | |
5830445C04Rik; C21orf127; HemK methyltransferase family member 2; HEMK2; m.HsaHemK2P; Methylarsonite methyltransferase N6AMT1; Methyltransferase N6AMT1; methyltransferase N6AMT1; hemK methyltransferase family member 2; M-N6DNA MTase A; MTQ2; N(6)-adenine-specific DNA methyltransferase 1; N(6)-adenine-specific DNA methyltransferase 1-like; N-6 adenine-specific DNA methyltransferase 1; N-6 adenine-specific DNA methyltransferase 1 (putative); N6AMT; N6amt1; N6-DNA methyltransferase A; N6-DNA-methyltransferase; PRED28; Protein N(5)-glutamine methyltransferase; RGD1311843 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
288309, 29104, 67768 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.92 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q9Y5N5 | |
N6AMT1 | |
A synthetic peptide corresponding to a sequence within amino acids 1-186 of human N6AMT1/HEMK2 (NP_8774263). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.