Learn More
N6AMT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 162111AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-0794
Add to Cart
Description
Description
Heterodimeric methyltransferase that catalyzes N5-methylation of ETF1 on ′Gln-185′, using S-adenosyl L-methionine as methyl donor. ETF1 needs to be complexed to ERF3 in its GTP-bound form to be efficiently methylated. May play a role in the modulation of arsenic-induced toxicity. May be involved in the conversion of monomethylarsonous acid (3+) into the less toxic dimethylarsonic acid.
Specifications
Specifications
N6AMT1
Polyclonal
Unconjugated
N6AMT1
C21orf127, HEMK2, MTQ2, N6AMT, N6AMT1, PRED28
Rabbit
Antigen Affinity Chromatography
RUO
288309, 29104, 67768
-20°C
Liquid
Western Blot
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
N6AMT1, Q9Y5N5
N6amt1
N6AMT1 Fusion Protein Ag9204
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions