Learn More
3M™ N95 Particulate Welding Respirators
N95 respirators with Cool Flow™ valve are designed to provide protection for applications such as welding or soldering where metal fumes may be present.
Supplier:  3M™ 7000002097
 View more versions of this product
Catalog No. 19-037-811
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Alert:
Do not exceed maximum use concentration established by regulatory standards. If respirator becomes damaged, soiled, or breathing becomes difficult, leave the contaminated area and dispose of the respirator. Before each use of the product, please inspect for any rips, damage, tears, etc. These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Description
Description
  • NIOSH approved N95 rating
  • Adjustable M-noseclip
  • Flame resistant per modified ASTM D2859
  • Recommended for use up to 10X the OSHA PEL or applicable occupational exposure limits, whichever is lower
  • For use when welding, torch cutting, soldering, brazing and working with molten metals
  • Cool Flow™ exhalation valve
Specifications
Specifications
N - Not Resistant to Oil
ASTM D2859
OSHA 1910.134
Cup/Molded
N95
Particulate Welding Respirator
Yes
Welding, torch cutting, soldering, brazing and working with molten metals
Braided Polyisoprene, Cotton/Polyester
Aluminum
None
Non-sterile
Adjustable Buckle Straps
10
NIOSH 42 CFR 84, N95, approval number: TC-84A-0454
White
Single-use
Polypropylene, Polyester
Standard
95%
Yes
8512
Standard
80/Cs.
No
No
Recommendations
Recommendations