Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NA-XTD™ Influenza Neuraminidase Assay Kit Non-distribution product as customer accommodation.

Next-generation chemiluminescence-based assay for long-lived emission kinetics for quantitation of influenza neuraminidase inhibitor resistance. Thermo Scientific NA-XTD Influenza Neuraminidase Assay Kit can be used for virus quantitation in media samples

$1496.00 - $1540.00

Specifications

For Use With (Application) Assessing and researching drug resistance of influenza. Screening for new neuraminidase inhibitors and quantitating viral neuraminidase activity for vaccine development
Format 96-well plate
Storage Requirements Store all components at 2° to 8°C.
View More Specs

Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
44-575-34 Non-distribution product as customer accommodation. Green Product View Documents Invitrogen™
4457534
NA-XTD Influenza Neuraminidase Assay Reagent Set Each for $1,496.00
Add to cart
 
44-575-35 Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only. Green Product View Documents Invitrogen™
4457535
NA-XTD Influenza Neuraminidase Assay Kit Each for $1,540.00
Add to cart
 
Description

Description

The NA-XTD kit includes detection reagents and microplates, eliminating the need for luminometers equipped with a reagent injector, and improving ease-of-use. This kit also includes complete assay protocols for quantitating sensitivity to neuraminidase inhibitors in various influenza virus isolates including: human influenza types A and B, A/H1N1 pandemic, avian, equine and porcine viruses.
 
  • Superior performance - improved, long-lived light emission kinetics and read-time flexibility
  • High sensitivity - high assay signal-to-noise, detection of low-virus concentrations, and wide assay dynamic range
  • Simple instrumentation requirement - no reagent injectors or luminometer needed
  • Multiple applications - neuraminidase inhibitor IC50 assays and cell-based virus quantitation
Long-Lived Light Emission Kinetics and Read-Time Flexibility
The NA-XTD assay yields much longer-lived light emission kinetics than does the NA-Star assay, eliminating the need for luminometer instrumentation with reagent injectors and enabling read-time flexibility and batch-mode processing of assay plates. NA-XTD assay signal half-life is approximately 2 hours, longer than the 5-10 minute half-life of the NA-Star assay signal. IC-50 values determined from data collected immediately or up to 3 hours following addition of NA-XTD Accelerator solution are identical.
 
High Sensitivity
The NA-XTD chemiluminescent assay provides higher detection sensitivity (better low-end detection limit), higher assay signal-to-noise ratio, and wider assay dynamic range than fluorescent assays with the MUNANA substrate. The NA-XTD assay also typically demonstrates slightly higher signal-to-noise than the NA-Star assay. The NA-XTD assay provides 2 to 50-fold higher sensitivity by signal-to-noise ratio than MUNANA-based fluorescence assays, depending on the virus isolate. The NA-XTD assay provides a dynamic range of detection of 3 to 4 orders of magnitude of neuraminidase enzyme concentration, compared to 2 to 3 orders of magnitude range with fluorescent MUNANA assays. The wide chemiluminescent neuraminidase assay range enables determination of IC-50 values over a wide range of virus concentrations, eliminating the need to titer virus prior to performing IC50 determination assays.
 
Simple Instrumentation Requirement
Optimally, a small sample of culture media is removed and assayed directly with NA-XTD reagents, permitting multiple samples to be assayed over time.
 
Specifications
  • Detection Method: Chemiluminescent
  • For Use With Equipment: Luminometer (Microplate)
  • Form: Liquid
  • Format: 96-well plate
  • High Throughput Compatibility: High Throughput-Compatible
  • Readout: End Point
  • Substrate: NA-XTD
  • Substrate Properties: Chemical Substrate
  • Substrate Type: Neuraminidase Substrate
  • Target Enzyme: Neuraminidase, Sialidase

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Assessing and researching drug resistance of influenza. Screening for new neuraminidase inhibitors and quantitating viral neuraminidase activity for vaccine development
96-well plate
Store all components at 2° to 8°C.

For Research Use Only. Not for use in diagnostics procedures.

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit