Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NAA20 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112622
Description
NAA20 Polyclonal Antibody for Western Blot, IP, ELISA
Catalytic subunit of the NatB complex which catalyzes acetylation of the N-terminal methionine residues of peptides beginning with Met-Asp, Met-Glu, Met-Asn and Met-Gln. Proteins with cell cycle functions are overrepresented in the pool of NatB substrates. Required for maintaining the structure and function of actomyosin fibers and for proper cellular migration.
Specifications
NAA20 | |
Polyclonal | |
Unconjugated | |
NAA20 | |
1500004D14Rik; 2900026I01Rik; AU041458; D2Ertd186e; dJ1002M8.1; Methionine N-acetyltransferase; N(alpha)-acetyltransferase 20, NatB catalytic subunit; NAA20; N-acetyltransferase 3 homolog; N-acetyltransferase 5; N-acetyltransferase 5 (ARD1 homolog, S. cerevisiae); N-acetyltransferase 5 (GCN5-related, putative); N-acetyltransferase 5, ARD1 subunit (arrest-defective 1, S. cerevisiae, homolog); N-alpha-acetyltransferase 20; N-alpha-acetyltransferase 20, NatB catalytic subunit; NAT3; NAT3P; NAT5; NAT5P; NatB catalytic subunit; natB complex subunit NAT5; N-terminal acetyltransferase B complex catalytic subunit NAA20; N-terminal acetyltransferase B complex catalytic subunit NAT5; N-terminal acetyltransferase complex ARD1 subunit | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
362228, 51126 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunoprecipitation, Western Blot | |
0.89 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P61599 | |
NAA20 | |
Recombinant Human N-alpha-acetyltransferase 20 protein (49-178AA). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction