Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAA30 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA563495
Description
Immunogen sequence: SLKSKVLSVA EVAATTATPD GGPRATATKG AGVHSGERPP HSLSSNARTA VPSPVEAAAA SDPAAARNGL A Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 62%.
Catalytic subunit of the N-terminal acetyltransferase C (NatC) complex. Catalyzes acetylation of the N-terminal methionine residues of peptides beginning with Met-Leu-Ala and Met-Leu-Gly. Necessary for the lysosomal localization and function of ARL8B sugeesting that ARL8B is a NatC substrate.Specifications
NAA30 | |
Polyclonal | |
Unconjugated | |
NAA30 | |
4930487N19Rik; 5730533P17Rik; AI447922; AW322455; C14orf35; MAK3; Mak3p; N(alpha)-acetyltransferase 30, NatC catalytic subunit; NAA30; N-acetyltransferase 12; N-acetyltransferase 12 (GCN5-related, putative); N-acetyltransferase MAK3 homolog; Nalpha acetyltransferase 30; N-alpha-acetyltransferase 30; N-alpha-acetyltransferase 30, NatC catalytic subunit; NAT12; NAT12P; NatC catalytic subunit; putative N-acetyltransferase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
122830 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q147X3 | |
NAA30 | |
Recombinant protein corresponding to Human NAA30 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?
Provide Content Correction