Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NAA40 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5140084
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: KSSKAKEKKQ KRLEERAAMD AVCAKVDAAN RLGDPLEAFP VFKKYDRNGL NVSIECKRVS GLEPATVDWA FD.
N-terminal acetylation is one of the most common protein modifications in eukaryotes, occurring on approximately 57% and 84% on yeast and human proteins respectively. There are several N-terminal acetylating enzyme complexes (NatA - NatE). Unlike the other complexes, NatD is composed of a single protein, NAT11, and has recently been described to acetylate the Serine N-termini of histones H2A and H4 in yeast. The role these modifications play is unknown; yeast that do not express NAT11 grow at normal rates and have no observable phenotypes. The role of the human homolog is likewise unknown.
Specifications
NAA40 | |
Polyclonal | |
Unconjugated | |
NAA40 | |
4931433E08Rik; AU023197; N(alpha)-acetyltransferase 40, NatD catalytic subunit; N(alpha)-acetyltransferase 40, NatD catalytic subunit, homolog; N(alpha)-acetyltransferase 40, NatD catalytic subunit, homolog (S. cerevisiae); Naa40; N-acetyltransferase 11; N-acetyltransferase 11 (GCN5-related, putative); N-alpha acetyl transferase 40; Nalpha acetyltransferase 40; N-alpha-acetyltransferase 40; N-alpha-acetyltransferase 40, NatD catalytic subunit; N-alpha-acetyltransferase D; NAT11; NatD; natD catalytic subunit; PATT1; Protein acetyltransferase 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79829 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86UY6 | |
NAA40 | |
Recombinant protein corresponding to Human NAA40. Recombinant protein control fragment (Product #RP-108926). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction